Orders LinkContactUsLinkComponent
Instrument trays
Instrument trays
400×270×10 mm
Catalog # USBE4764
Supplier:   USBECK Laborgeräte
Instrument trays
Catalog # USBE4764
Instrument trays
Instrument trays
400×270×10 mm
Price
53,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Easy to clean
Instrument trays with rounded corner.
Specifications
  • Seq:
    03
  • W×D×H:
    400×270×10 mm