Instrument trays
180×120×10 mm
Catalog # USBE4760
Supplier: USBECK Laborgeräte
Jump to:
Product Details & Documents
Stainless steel.
- Excellent chemical resistance
- Excellent temperature resistance
- Easy to clean
Instrument trays with rounded corner.
Specifications
- Seq:01
- W×D×H:180×120×10 mm