ContactUsLinkComponent
TSHR - Human shRNA lentiviral particles (4 unique 29mer target-specific shRNA, 1 scramble control), 0.5 ml each, >10
Catalog # ORIGTL308613V
Supplier:  OriGene
TSHR - Human shRNA lentiviral particles (4 unique 29mer target-specific shRNA, 1 scramble control), 0.5 ml each, >10
Catalog # ORIGTL308613V
Supplier:  OriGene