Orders LinkContactUsLinkComponent
Accessories
Accessories
Telescoping aspirating tube, FEP, individually adjustable length 170 mm-330 mm, for Varispenser 2(x) with nominal volume of 25 ml, 50 ml or 100 ml
Catalog # EPPE4966507.000
Supplier:   EPPENDORF
Accessories
Catalog # EPPE4966507.000
Supplier:  EPPENDORF
Accessories
Accessories
Telescoping aspirating tube, FEP, individually adjustable length 170 mm-330 mm, for Varispenser 2(x) with nominal volume of 25 ml, 50 ml or 100 ml
Price
37,40 €
Log in for availability
1 * 1 SET
Product Details & Documents
Specifications
  • Description:
    Telescoping aspirating tube, FEP, individually adjustable length 170 mm-330 mm, for Varispenser 2(x) with nominal volume of 25 ml, 50 ml or 100 ml