Orders LinkContactUsLinkComponent
Capillary installation gauges
Capillary installation gauges
Capillary installation gauge for "Bruker/Varian GCs for use with 1/16" ferrules"
Catalog # REST22335
Supplier:   Restek
Capillary installation gauges
Catalog # REST22335
Supplier:  Restek
Capillary installation gauges
Capillary installation gauges
Capillary installation gauge for "Bruker/Varian GCs for use with 1/16" ferrules"
Price
121,00 €
Log in for availability
1 * 1 items
Product Details & Documents
These capillary installation gauges for Agilent GCs are made from high-quality stainless steel.
  • Prevents crushed column ends
  • Seats graphite ferrules onto column for consistent installations
Specifications
  • Description:
    Capillary installation gauge for "Bruker/Varian GCs for use with 1/16" ferrules"