[2.2]-Paracyclophane 99%
Glass bottle, 25 g
Catalog # ACRO291600250
Supplier: Thermo Fisher Scientific
[2.2]-Paracyclophane 99%
Catalog # ACRO291600250
Supplier: Thermo Fisher Scientific
CAS Number: 1633-22-3
Jump to:
Product Details & Documents
Synonyms: Tricyclo[8,2,2,24,7]hexadeca-4,6,10,12,13,15-hexaene, Tricyclo[8.2.2.2(4,7)]hexadeca-1(12),4,6,10,13,15-hexaene
- CAS Number:1633-22-3
- Formula:C₁₆H₁₆
- MDL Number:MFCD00003707
- Molecular Weight:208.3 g/mol
- Melting Pt:285…288 °C
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:White to off-white Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:285 to 288 °C
- HPLC:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:25 g



