Tobramycin 95+%
Glass bottle, 5 g
Catalog # ACRO455430050
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: (2S,3R,4S,5S,6R)-4-Amino-2-(((1S,2S,3R,4S,6R)-4,6-diamino-3-(((2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxytetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,5-diol
- CAS Number:32986-56-4
- Formula:C₁₈H₃₇N₅O₉
- MDL Number:MFCD00077885
- Molecular Weight:467.51 g/mol
- Storage Temperature:Ambient
Specifications
Specification Test Results
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Water:≤8 %
- Titration with HClO4:≥95.0 % (on dry substance)
Specifications
- Pack type:Glass bottle
- Pk:5 g



