Order Entry
Export
Orders LinkContactUsLinkComponent
Tobramycin 95+%

Tobramycin 95+%

Glass bottle, 1 g
Catalog # ACRO455430010
Tobramycin 95+%
Catalog # ACRO455430010
CAS Number:  32986-56-4
Tobramycin 95+%
Tobramycin 95+%

Glass bottle, 1 g

CAS Number:  32986-56-4
Price
55,20 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  (2S,3R,4S,5S,6R)-4-Amino-2-(((1S,2S,3R,4S,6R)-4,6-diamino-3-(((2R,3R,5S,6R)-3-amino-6-(aminomethyl)-5-hydroxytetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,5-diol
  • CAS Number:32986-56-4
  • Formula:C₁₈H₃₇N₅O₉
  • MDL Number:MFCD00077885
  • Molecular Weight:467.51 g/mol
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Appearance:
    White to light yellow Powder
  • Infrared spectrum:
    Conforms
  • Water:
    ≤8 %
  • Titration with HClO4:
    ≥95.0 % (on dry substance)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g

Specifications