Streptomycin sulphate
Glass bottle, 25 g
Catalog # ACRO455340250
Supplier: Thermo Fisher Scientific
Streptomycin sulphate
Catalog # ACRO455340250
Supplier: Thermo Fisher Scientific
CAS Number: 3810-74-0
Jump to:
Product Details & Documents
- CAS Number:3810-74-0
- Formula:C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
- MDL Number:MFCD00037023
- Molecular Weight:1457.4 g/mol
- Storage Temperature:Refrigerator
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Loss on drying:≤7 %
- Sulfate (SO4):18.0 to 21.5
Specifications
- Pack type:Glass bottle
- Pk:25 g



