Orders LinkContactUsLinkComponent
Streptomycin sulphate
Glass bottle, 5 g
  ACRO455340050
Streptomycin sulphate
  ACRO455340050
 :  3810-74-0
Streptomycin sulphate
Streptomycin sulphate
Glass bottle, 5 g
 :  3810-74-0
32,10 €
1 * 5 g
  •  :3810-74-0
  •  :C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
  •  :MFCD00037023
  •  :1457.4 g/mol
  •  :Refrigerator
  • Appearance:
    White to off-white Powder
  • Loss on drying:
    ≤7 %
  • Sulfate (SO4):
    18.0 to 21.5
  • Pack type:
    Glass bottle
  • Pk:
    5 g