Order Entry
Export
Orders LinkContactUsLinkComponent
Gentamicin sulphate

Gentamicin sulphate

Glass bottle, 1 g
Catalog # ACRO455310010
Health
Gentamicin sulphate
Catalog # ACRO455310010
CAS Number:  1405-41-0
Gentamicin sulphate
Gentamicin sulphate

Glass bottle, 1 g

CAS Number:  1405-41-0
Price
37,30 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C
  • CAS Number:1405-41-0
  • MDL Number:MFCD00270181
  • Melting Pt:218…237 °C
  • Storage Temperature:Refrigerator
Specifications

Specification Test Results

  • Appearance:
    White to yellow Powder
  • Infrared spectrum:
    Conforms
  • Assay:
    ≥590 µg/mg (On dried basis)
  • Loss on drying:
    ≤18.0 % (Vacuum, 110°C, 3h)
  • Sulfated ash:
    ≤1.0 %
  • pH:
    3.5 to 5.5 (c=4, water)
  • Specific optical rotation:
    +107° to +121° (20°C, 589 nm) (c=1, water)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g

Specifications