Ethanolamine ACS
Glass bottle, 250 mL
ACRO451762500
: 2-Aminoethanol, Monoethanolamine
- :141-43-5
- :NH₂CH₂CH₂OH
- :MFCD00008183
- :61.08 g/mol
- :170 °C (760 mmHg)
- :10.3 °C
- :1.012 g/cm³
- :92 °C
- :Ambient
- :2491
- :8,III
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.30 %
- GC:≥99.0 %
- Color scale:≤15 APHA
- Iron (Fe):≤5 ppm
- Heavy metals (as Pb):≤5 ppm
- Pack type:Glass bottle
- Pk:250 mL



