Order Entry
Export
Orders LinkContactUsLinkComponent
 

Ethanolamine ACS

Glass bottle, 2,5 L
Catalog # ACRO451760025
Irritant
Corrosive
Ethanolamine ACS
Catalog # ACRO451760025
CAS Number:  141-43-5
Ethanolamine ACS
Ethanolamine ACS

Glass bottle, 2,5 L

CAS Number:  141-43-5
Price
140,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  2-Aminoethanol, Monoethanolamine
  • CAS Number:141-43-5
  • Formula:NH₂CH₂CH₂OH
  • MDL Number:MFCD00008183
  • Molecular Weight:61.08 g/mol
  • Boiling Pt:170 °C (760 mmHg)
  • Melting Pt:10.3 °C
  • Density:1.012 g/cm³
  • Flash Pt:92 °C
  • Storage Temperature:Ambient
  • UN:2491
  • ADR:8,III
Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.30 %
  • GC:
    ≥99.0 %
  • Color scale:
    ≤15 APHA
  • Iron (Fe):
    ≤5 ppm
  • Heavy metals (as Pb):
    ≤5 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications