Order Entry
Export
Orders LinkContactUsLinkComponent
 

Quartz, fine wool 4 µm

Glass bottle, 50 g
Catalog # ACRO451040500
Health
Quartz, fine wool 4 µm
Catalog # ACRO451040500
CAS Number:  14808-60-7
Quartz, fine wool 4 µm
Quartz, fine wool 4 µm

Glass bottle, 50 g

CAS Number:  14808-60-7
Price
181,00 €
Log in for availability
1 * 50 g
Product Details & Documents
Synonyms:  Silicium dioxide, Silicium (IV) oxide, Silicon dioxide, Sand, white quartz
  • CAS Number:14808-60-7
  • Formula:SiO₂
  • MDL Number:MFCD00217788
  • Molecular Weight:60.08 g/mol
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Appearance:
    White Fibers

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 g

Specifications