2'-Deoxycytidine hydrochloride 98%
Glass bottle, 5 g
Catalog # ACRO458510050
Supplier: Thermo Fisher Scientific
2'-Deoxycytidine hydrochloride 98%
Catalog # ACRO458510050
Supplier: Thermo Fisher Scientific
CAS Number: 3992-42-5
Jump to:
Product Details & Documents
Synonyms: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride
- CAS Number:3992-42-5
- Formula:C₉H₁₄ClN₃O₄
- MDL Number:MFCD00012840
- Molecular Weight:263.68 g/mol
- Storage Temperature:Freezer
Specifications
Specification Test Results
- Appearance:Off-white to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Water:≤8 %
- HPLC:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:5 g



