2,2':6',2''-Terpyridine 96%
Glass bottle, 5 g
Catalog # ACRO215630050
Supplier: Thermo Fisher Scientific
2,2':6',2''-Terpyridine 96%
Catalog # ACRO215630050
Supplier: Thermo Fisher Scientific
CAS Number: 1148-79-4
Jump to:
Product Details & Documents
Synonyms: TERPY
- CAS Number:1148-79-4
- Formula:C₁₅H₁₁N₃
- MDL Number:MFCD00006213
- Molecular Weight:233.27 g/mol
- Boiling Pt:370 °C
- Melting Pt:89…91 °C
- Storage Temperature:Ambient
- UN:2811
- ADR:6.1,I
Specifications
Specification Test Results
- Appearance:Cream to yellow to brown Crystalline powder
- Infrared spectrum:Conforms
- Melting point:89 to 95 °C
- GC:≥95 %
Specifications
- Pack type:Glass bottle
- Pk:5 g



