Mannose triflate 98%
Glass bottle, 100 mg
Catalog # ACRO461341000
Supplier: Thermo Fisher Scientific
Mannose triflate 98%
Catalog # ACRO461341000
Supplier: Thermo Fisher Scientific
CAS Number: 92051-23-5
Jump to:
Product Details & Documents
Synonyms: (2S,3S,4S,5R,6R)-6-(Acetoxymethyl)-3-(((trifluoromethyl)sulphonyl)oxy)tetrahydro-2H-pyran-2,4,5-triyl triacetate, 1,3,4,6-Tetra-O-acetyl-2-O-(trifluoromethanesulphonyl)-β-D-mannopyranose, TATM
- CAS Number:92051-23-5
- Formula:C₁₅H₁₉F₃O₁₂S
- MDL Number:MFCD00012353
- Molecular Weight:480.36 g/mol
- Melting Pt:118…122 °C
- Storage Temperature:Refrigerator
Specifications
Specification Test Results
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Melting point:118 to 122 °C
- Solubility:Clear colorless to slightly yellow
- HPLC:≥97.5 % (ELSD)
- Specific optical rotation:-12° to -19° (20 °C, 589 nm) (1 % in CH3CN)
Specifications
- Pack type:Glass bottle
- Pk:100 mg



