Order Entry
Export
Orders LinkContactUsLinkComponent
Estriol 97%

Estriol 97%

Glass bottle, 1 g
Catalog # ACRO460490010
Health
Estriol 97%
Catalog # ACRO460490010
CAS Number:  50-27-1
Estriol 97%
Estriol 97%

Glass bottle, 1 g

CAS Number:  50-27-1
Price
101,00 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:50-27-1
  • Formula:C₁₈H₂₄O₃
  • MDL Number:MFCD00003691
  • Melting Pt:280…282 °C
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤0.5 %
  • HPLC:
    ≥96.0 %
  • Specific optical rotation:
    +54° to +62° (20°C, 589 nm) (0.4 % in Dioxane)

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g

Specifications