Orders LinkContactUsLinkComponent
KORASILON™ pastes
KORASILON™ pastes
KORASILON™ paste, low viscosity
Catalog # SIAL769711-1EA
Supplier:   Merck
KORASILON™ pastes
Catalog # SIAL769711-1EA
Supplier:  Merck
KORASILON™ pastes
KORASILON™ paste, low viscosity
Price
48,40 €
Log in for availability
1 * 1 items
Product Details & Documents
The pastes are excellently suitable as glide-, lubricant- and release agent for e. g. vales, joints and gaskets, ground joints, cables and wires. They are frequently also used as assembly aid as well as protection- and sealing aid. Furthermore, we recommend the pastes for maintaining rubber seals. In electronics, they find their use for the assembly of insulators and switchgears amongst others.
  • Stable at temperatures ranging from -50 °C to more than 300 °C
  • Exceptional hydrophobic and practically insoluble in water
  • Show thixotropic behavior
Specifications
  • Description:
    KORASILON™ paste, low viscosity
  • Seq:
    01