Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CIPROFLOXACIN 5MCG 5VIALS X 100DISCS 1 * 500 items
Catalog # HIMESD060-5VL
Supplier:
HIMEDIA
Expand image
CIPROFLOXACIN 5MCG 5VIALS X 100DISCS 1 * 500 items
Catalog # HIMESD060-5VL
Supplier:
HIMEDIA