Electronic overhead stirrers, MICROSTAR control
MICROSTAR 7.5 control overhead stirrer
Catalog # 441-0464
Supplier: IKA
Jump to:
Product Details & Documents
These overhead stirrers are ideal for special applications. Combining high performance with excellence, the units require minimum space.
- Fast, responsive display for maximum visibility and chemical resistance
- Clear display shows all essential information at a glance
- Integrated timer/counter for the control of kinetic sensitive reactions and reminders
- USB interface for documenting parameters using labworldsoft® or installing firmware updates
- Intuitive and simple handling; touch-sensitive surface for long service life
- Temperature reading is shown on display
- Chemical resistant housing is enclosed in hardened glass
- Key lock function
Microprocessor-controlled speed monitoring provides constant rotational speed, even with changes in viscosity. The state-of-art vibration sensors detect deviations from permissible thresholds and automatically stop the process.
Temperature measurement range: −10 to +350 °C.
: IP protection class according to DIN EN 60529: IP 54
Specifications
- Description:MICROSTAR 7.5 control overhead stirrer
- Seq:01
- Chuck range:0,5 - 8,0 mm
- Max. torque at chuck:7,5 Ncm
- Power - motor input/output:32/22 W
- Max. viscosity:4000 mPas
- Weight:1,18 kg
- Plug in coupling Ø:32/22 mm
- Permissible relative humidity:80%
- Cat. No.:441-0464
- Model:MICROSTAR 7.5 control
- W×D×H:60×110×123 mm
- Max. stirring capacity H₂O:5 L
- Speed range I:30 - 2000 min⁻¹
- Permissible ambient temperature:5…40 °C
Accessories
Product Information
- Catalog #AvailabilityPrice
- H 71.45 Temperature sensor 450 mm, PTFE coated, Ø 8 mmH 71.45 Temperature sensor 450 mm, PTFE coated, Ø 8 mmIKAA20118833Log in for availability1 * 1 items - 717,00 €
Product Information
- Catalog #AvailabilityPrice
- Extension cable, separates casing from the sensor, avoiding damage from vapours released by the mediumExtension cable, separates casing from the sensor, avoiding damage from vapours released by the medium620-8104Log in for availability1 * 1 items - 248,00 €
Product Information
- Catalog #Stirrer ØShaft ØAvailabilityPrice
- Dissolver stirrer
Product Information
- Catalog #AvailabilityPrice
- IKA [scale] Weighing stand, capacity 5 to 6000 gIKA [scale] Weighing stand, capacity 5 to 6000 gIKAA25004318Log in for availability1 * 1 items - 1 010,00 €
Product Information
- Catalog #Stirrer ØShaft ØAvailabilityPrice
- Centrifugal stirrer
Product Information
- Catalog #ForAvailabilityPrice
- R 60.1 Keyless chuckR 60.1 Keyless chuckIKAA25007821use with MINISTAR 20/40/80 digital and control overhead stirrersLog in for availability1 * 1 items - 99,50 €
Product Information
- Catalog #AvailabilityPrice
- Strap clamp for securing vessels, stand Ø 25 - 36 mm, vessel Ø 40 - 300 mmStrap clamp for securing vessels, stand Ø 25 - 36 mm, vessel Ø 40 - 300 mm431-2944Log in for availability1 * 1 items - 446,00 €
Product Information
- Catalog #Stirrer ØShaft ØAvailabilityPrice
- Dissolver stirrer
Product Information
- Catalog #ForAvailabilityPrice
- Labworldsoft® 6 StarterLabworldsoft® 6 StarterIKAA20019397Small configurations with up to 3 devices can be controlled at the same timeLog in for availability1 * 1 items - 6 500,00 €
Product Information
- Catalog #Stirrer ØShaft ØAvailabilityPrice
- 3-blade stirrer
Product Information
- Catalog #AvailabilityPrice
- Glass coated sensor for use with ETS-D5 and VT-5 contact thermometersGlass coated sensor for use with ETS-D5 and VT-5 contact thermometers442-0316Log in for availability1 * 1 items - 357,00 €
Product Information
- Catalog #ForTypeAvailabilityPrice
- Plate stand, 200×316 mm, rod 16 mm Ø with height 1000 mmPlate stand, 200×316 mm, rod 16 mm Ø with height 1000 mm441-2129T 18 basic, T 25 digitalPlate stand R 1827Log in for availability1 * 1 items - 418,00 €
Product Information
- Catalog #Stirrer ØShaft ØAvailabilityPrice
- 4-blade stirrer
Product Information
- Catalog #ForTypeAvailabilityPrice
- Plate stand, 200×316 mm, rod 16 mm Ø with height 800 mmPlate stand, 200×316 mm, rod 16 mm Ø with height 800 mm431-2901T 18 basic, T 25 digitalPlate stand R 1826, plate 200x316 mm, rod 800 mmLog in for availability1 * 1 items - 392,00 €
Product Information
- Catalog #AvailabilityPrice
- Plate stand R 1825, 200×316 mm, rod 16 mm Ø with height 560 mmPlate stand R 1825, 200×316 mm, rod 16 mm Ø with height 560 mm431-1047Log in for availability1 * 1 items - 355,00 €
Product Information
- Catalog #AvailabilityPrice
- Flexible coupling for applications using glass stirring rods Ø 6 - 10 mm, max. torque 10 NcmFlexible coupling for applications using glass stirring rods Ø 6 - 10 mm, max. torque 10 Ncm441-9213Log in for availability1 * 1 items - 377,00 €
Product Information
- Catalog #Stirrer ØShaft ØAvailabilityPrice
- Spiral stirrer
Product Information
- Catalog #AvailabilityPrice
- Bosshead clamp, 25 - 36 mm Ø stand, 5 - 21 mm Ø extension armBosshead clamp, 25 - 36 mm Ø stand, 5 - 21 mm Ø extension arm431-0004Log in for availability1 * 1 items - 147,00 €
Product Information
- Catalog #Stirrer ØShaft ØAvailabilityPrice
- 3-blade stirrer
Product Information
- Catalog #AvailabilityPrice
- Pneumatic telescopic stand, 460×420 mm, rod 34 mm Ø with height 620 - 1010 mmPneumatic telescopic stand, 460×420 mm, rod 34 mm Ø with height 620 - 1010 mm431-2912Log in for availability1 * 1 items - 1 580,00 €
Product Information
- Catalog #AvailabilityPrice
- R 1346 (PTFE-coated) Propeller stirrer, 4-bladed, 350 mm shaft length, Stirrer Ø 58 mm, Shaft Ø: 8 mm (uncoated part Ø: 6 mm)R 1346 (PTFE-coated) Propeller stirrer, 4-bladed, 350 mm shaft length, Stirrer Ø 58 mm, Shaft Ø: 8 mm (uncoated part Ø: 6 mm)IKAA25006431Log in for availability1 * 1 items - 305,00 €
Product Information
- Catalog #AvailabilityPrice
- Stirring shaft protector
Product Information
- Catalog #Stirrer ØShaft ØAvailabilityPrice
- Centrifugal stirrer
Product Information
- Catalog #Stirrer ØShaft ØAvailabilityPrice
- Anchor stirrer
Product Information
- Catalog #Stirrer ØShaft ØAvailabilityPrice
- 4-blade stirrer
Product Information
- Catalog #AvailabilityPrice
- Stainless steel screw type stirrer
Product Information
- Catalog #AvailabilityPrice
- Spare stainless steel sensor for use with ETS-D5Spare stainless steel sensor for use with ETS-D5442-0252Log in for availability1 * 1 items - 245,00 €
Product Information
- Catalog #AvailabilityPrice
- H-Stand, 460×420 mm, rod 34 mm Ø with height 1010 mmH-Stand, 460×420 mm, rod 34 mm Ø with height 1010 mm431-9114Log in for availability1 * 1 items - 851,00 €
Product Information
- Catalog #Stirrer ØShaft ØAvailabilityPrice
- Anchor stirrer
Product Information
- Catalog #ForTypeAvailabilityPrice
- Strap clamp for securing vessels, stand Ø 8 - 16 mm, vessel Ø 40 - 300 mmStrap clamp for securing vessels, stand Ø 8 - 16 mm, vessel Ø 40 - 300 mm431-2943T 18 basic, T 25 digitalClamp device RH 3Log in for availability1 * 1 items - 363,00 €
Product Information
- Catalog #ForAvailabilityPrice
- Labworldsoft® 6 Pro, with access to the entire device libraryLabworldsoft® 6 Pro, with access to the entire device libraryIKAA20017366All configurations with any number of devices can be controlled at the same timeLog in for availability1 * 1 items - 14 820,00 €
Product Information
- Catalog #Stirrer ØShaft ØAvailabilityPrice
- Paddle stirrer
Product Information
- Catalog #AvailabilityPrice
- Beaker cap, BC 1000, silicone, for 600 and 1000 ml beakersBeaker cap, BC 1000, silicone, for 600 and 1000 ml beakersIKAA20003417Log in for availability1 * 1 items - 22,50 €
Product Information
- Catalog #AvailabilityPrice
- H 71.32 Temperature sensor 320 mm, PTFE coated, Ø 8 mmH 71.32 Temperature sensor 320 mm, PTFE coated, Ø 8 mmIKAA20117772Log in for availability1 * 1 items - 662,00 €
Product Information
- Catalog #AvailabilityPrice
- Support holder for stirring shaft
Product Information
- Catalog #Stirrer ØShaft ØAvailabilityPrice
- 3-blade stirrer (PTFE)
Product Information
- Catalog #AvailabilityPrice
- Spare stainless steel paddle stirrer
Product Information
- Catalog #AvailabilityPrice
- Temperature sensor, coated
Product Information
- Catalog #Stirrer ØShaft ØAvailabilityPrice
- Turbine stirrer
Product Information
- Catalog #Stirrer ØShaft ØAvailabilityPrice
- Turbine stirrer
Product Information
- Catalog #AvailabilityPrice
- Radial stirrer, R 3004, shaft (Ø×L) 8,0×359 mm, stirrer Ø 30 mmRadial stirrer, R 3004, shaft (Ø×L) 8,0×359 mm, stirrer Ø 30 mmIKAA20001206Log in for availability1 * 1 items - 363,00 €
Product Information
- Catalog #ForTypeAvailabilityPrice
- Bosshead clamp, 6 - 16 mm Ø stand, 6 - 16 mm Ø extension armBosshead clamp, 6 - 16 mm Ø stand, 6 - 16 mm Ø extension arm431-2923T 18 basic, T 25 digitalFixing clamp R 182Log in for availability1 * 1 items - 113,00 €