Sterilisation -Maximum Thermometer, DIN 12770, Capillary Yellow Round, +80+135C in 0.5C, Mercury, Immersion total, Length 200 mm, with flat button with safety cover, + Factory Test certificate calibrated at +120C, with Metal Protection 1 * 1 items
Catalog # SCNL6435280031P608
Supplier: Ludwig Schneider GmbH & CO. KG