Orders LinkContactUsLinkComponent
Connecting adapters, with parallel necks
Connecting adapters, with parallel necks
18,8/26 NS, 18,8/26 NS
Catalog # NEUB0900-10-191929
Connecting adapters, with parallel necks
Catalog # NEUB0900-10-191929
Connecting adapters, with parallel necks
18,8/26 NS, 18,8/26 NS
Price
85,60 €
Log in for availability
1 * 1 items
Product Details & Documents
Two necked adapters made of borosilicate glass 3.3
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Specifications
  • Cone size:
    18,8/26 NS
  • Socket size:
    18,8/26 NS
  • Seq:
    02