Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
15 (380mm) Straight 316 SS 1/8 (3.2mm) OD Cannula with SS Luer Lock 1 * 1 items
Catalog # QLAACANS15-SH
Supplier:
QUALITY LAB ACCESSORIES
Expand image
15 (380mm) Straight 316 SS 1/8 (3.2mm) OD Cannula with SS Luer Lock 1 * 1 items
Catalog # QLAACANS15-SH
Supplier:
QUALITY LAB ACCESSORIES