Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TN 552/1 GAPP - Temperature controller compact model, analog output, 1 switching point, measurin range -40C ... +120C 1 * 1 ST
Catalog # EGEEP71020
Supplier:
EGE ELKTRONIK SPEZIAL SENSOREN
Expand image
TN 552/1 GAPP - Temperature controller compact model, analog output, 1 switching point, measurin range -40C ... +120C 1 * 1 ST
Catalog # EGEEP71020
Supplier:
EGE ELKTRONIK SPEZIAL SENSOREN