Instrument trays
215×315×55 mm
Catalog # USBE4744
Supplier: USBECK Laborgeräte
Jump to:
Product Details & Documents
Made of stainless steel.
- Rounded corners
- With rim
- Very good chemical and temperature resistance
Specifications
- W×D×H:215×315×55 mm