Orders LinkContactUsLinkComponent
Coats, paint-tex® plus
Coats, paint-tex® plus
White, XL
Catalog # ZVGA30774-00
Coats, paint-tex® plus
Catalog # ZVGA30774-00
Coats, paint-tex® plus
Coats, paint-tex® plus
White, XL
Price
432,00 €
Log in for availability
1 * 50 items
Product Details & Documents
Suitable for longer use due to the breathable SMS material.
  • Features snap buttons
  • Collar and cuffs
  • Three pockets
Ideal for renovation work, in the remediation of contaminated sites and asbestos.
 : Cat. III/CE 1023/Type 5 + 6
Specifications
  • Clothing Size:
    XL
  • Colour:
    White
  • Seq:
    02
  • Cat. No.:
    ZVGA30774-00
  • Garment application:
    Industrial
  • Pockets:
    With pockets
  • Closure type:
    Press studs / snaps