Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
4-neck sulfonation flask, 350ml, 1x center neck NS29/32, 3x side necks NS14/23 1 * 1 items
Catalog # WITG0.851.035
Supplier:
witeg Labortechnik
Expand image
4-neck sulfonation flask, 350ml, 1x center neck NS29/32, 3x side necks NS14/23 1 * 1 items
Catalog # WITG0.851.035
Supplier:
witeg Labortechnik