Coats, paint-tex® plus
White, XXL
Catalog # ZVGA30775-00
Supplier: Zellstoff-Vertriebs-GmbH & Co. KG
Jump to:
Product Details & Documents
Suitable for longer use due to the breathable SMS material.
- Features snap buttons
- Collar and cuffs
- Three pockets
Ideal for renovation work, in the remediation of contaminated sites and asbestos.
: Cat. III/CE 1023/Type 5 + 6
Specifications
- Clothing Size:XXL
- Colour:White
- Seq:03
- Cat. No.:ZVGA30775-00
- Garment application:Industrial
- Pockets:With pockets
- Closure type:Press studs / snaps