ContactUsLinkComponent
TRANSFERRIN(1F10) MONOCLONAL ANTIBODY, ALEXA FLUOR® 488 CONJUGATED, HOST: MOUSE, CROSS REACTIVE SPECIES: HUMAN, APPLICATION: IF(IHC- P), CONCENTRATION: 1UG/UL, VOLUME: 100UL 1 * 100 µl
Catalog # BOSSBSM-2052M-A488
Supplier:  Bioss
TRANSFERRIN(1F10) MONOCLONAL ANTIBODY, ALEXA FLUOR® 488 CONJUGATED, HOST: MOUSE, CROSS REACTIVE SPECIES: HUMAN, APPLICATION: IF(IHC- P), CONCENTRATION: 1UG/UL, VOLUME: 100UL 1 * 100 µl
Catalog # BOSSBSM-2052M-A488
Supplier:  Bioss
Related Information