ContactUsLinkComponent
CD4 POLYCLONAL ANTIBODY, BIOTIN CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: HUMAN, APPLICATION: WB, IHC-P, CONCENTRATION: 1UG/ UL, VOLUME: 100UL 1 * 100 µl
Catalog # BOSSBS-4817R-BIOT
Supplier:  Bioss
CD4 POLYCLONAL ANTIBODY, BIOTIN CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: HUMAN, APPLICATION: WB, IHC-P, CONCENTRATION: 1UG/ UL, VOLUME: 100UL 1 * 100 µl
Catalog # BOSSBS-4817R-BIOT
Supplier:  Bioss
Related Information