ContactUsLinkComponent
SORCS1 POLYCLONAL ANTIBODY, BIOTIN CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: HUMAN, MOUSE, RAT, APPLICATION: WB, IHC-P, CONC ENTRATION: 1UG/UL, VOLUME: 100UL 1 * 100 µl
Catalog # BOSSBS-11330R-BIOT
Supplier:  Bioss
SORCS1 POLYCLONAL ANTIBODY, BIOTIN CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: HUMAN, MOUSE, RAT, APPLICATION: WB, IHC-P, CONC ENTRATION: 1UG/UL, VOLUME: 100UL 1 * 100 µl
Catalog # BOSSBS-11330R-BIOT
Supplier:  Bioss
Related Information