ContactUsLinkComponent
SHH POLYCLONAL ANTIBODY, ALEXA FLUOR® 594 CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: HUMAN, MOUSE, RAT, APPLICATION: IF(IHC-P ), CONCENTRATION: 1UG/UL, VOLUME: 100UL 1 * 100 µl
Catalog # BOSSBS-1544R-A594
Supplier:  Bioss
SHH POLYCLONAL ANTIBODY, ALEXA FLUOR® 594 CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: HUMAN, MOUSE, RAT, APPLICATION: IF(IHC-P ), CONCENTRATION: 1UG/UL, VOLUME: 100UL 1 * 100 µl
Catalog # BOSSBS-1544R-A594
Supplier:  Bioss
Related Information