ContactUsLinkComponent
KY/KYPHOSCOLIOSIS PEPTIDASE ANTIBODY, CY5.5 CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: HUMAN, MOUSE, RAT, APPLICATION: IF(IHC -P), CONCENTRATION: 1UG/UL, VOLUME: 100UL 1 * 100 µl
Catalog # BOSSBS-11174R-CY55
Supplier:  Bioss
KY/KYPHOSCOLIOSIS PEPTIDASE ANTIBODY, CY5.5 CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: HUMAN, MOUSE, RAT, APPLICATION: IF(IHC -P), CONCENTRATION: 1UG/UL, VOLUME: 100UL 1 * 100 µl
Catalog # BOSSBS-11174R-CY55
Supplier:  Bioss