ContactUsLinkComponent
EIF4EBP1 (SER65) POLYCLONAL ANTIBODY, ALEXA FLUOR® 594 CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: HUMAN, MOUSE, RAT, APPLICAT ION: IF(IHC-P), CONCENTRATION: 1UG/UL, VOLUME: 100UL 1 * 100 µl
Catalog # BOSSBS-3018R-A594
Supplier:  Bioss
EIF4EBP1 (SER65) POLYCLONAL ANTIBODY, ALEXA FLUOR® 594 CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: HUMAN, MOUSE, RAT, APPLICAT ION: IF(IHC-P), CONCENTRATION: 1UG/UL, VOLUME: 100UL 1 * 100 µl
Catalog # BOSSBS-3018R-A594
Supplier:  Bioss
Related Information