ContactUsLinkComponent
COX4 POLYCLONAL ANTIBODY, CY5 CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: HUMAN, MOUSE, RAT, APPLICATION: FCM, IF(ICC), CONCEN TRATION: 1UG/UL, VOLUME: 100UL 1 * 100 µl
Catalog # BOSSBS-10257R-CY5
Supplier:  Bioss
COX4 POLYCLONAL ANTIBODY, CY5 CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: HUMAN, MOUSE, RAT, APPLICATION: FCM, IF(ICC), CONCEN TRATION: 1UG/UL, VOLUME: 100UL 1 * 100 µl
Catalog # BOSSBS-10257R-CY5
Supplier:  Bioss
Related Information