ContactUsLinkComponent
C1ORF182 POLYCLONAL ANTIBODY, CY5.5 CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: HUMAN, APPLICATION: IF(IHC-P), CONCENTRATION: 1UG/UL, VOLUME: 100UL 1 * 100 µl
Catalog # BOSSBS-15043R-CY55
Supplier:  Bioss
C1ORF182 POLYCLONAL ANTIBODY, CY5.5 CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: HUMAN, APPLICATION: IF(IHC-P), CONCENTRATION: 1UG/UL, VOLUME: 100UL 1 * 100 µl
Catalog # BOSSBS-15043R-CY55
Supplier:  Bioss