Order Entry
Export
Orders LinkContactUsLinkComponent
 

VWR®, Beakers, Low Form, PP

250 ml
  213-0491
   Sustainable Packaging
VWR®, Beakers, Low Form, PP
  213-0491
   Sustainable Packaging
VWR®, Beakers, Low Form, PP
VWR®, Beakers, Low Form, PP

250 ml

 
12,30 €
   
1 * 6 items
 

PP, translucent.

  • Printed graduations
  • Can be stacked when empty to save space while storing
  • Withstand temperatures of 0 to 121 °C
  • Resin meets US FDA 21 CFR requirements

Ideal for general laboratory use. With spout and exceptionally 'easy to read' printed graduations.

 
  • Capacity:
    250 ml
  • Seq:
    04
  • Cat. No.:
    213-0491
  • Environmentally Preferable:
    Sustainable Packaging