Various replacement parts
Capillary, loop, 100 µl, 1100, 1120, 1220, G1313A, G1329A/B, 01078-87302, AGLS
Catalog # CTSA11-4059
Supplier: SCIENCIX
Jump to:
Product Details & Documents
Replacement parts for OEMs such as Agilent, Shimadzu and Waters chromotograpy systems. Equivalent to the corresponding OEM part.
Documents
CTSACTS-11459_TDS_EN
Datasheet
Specifications
- Description:Capillary, loop, 100 µl
- OEM manufacturer:AGLS
- OEM equivalent:01078-87302
- Model:1100, 1120, 1220, G1313A, G1329A/B