Order Entry
Export
Orders LinkContactUsLinkComponent
Human Recombinant FGF-18

Human Recombinant FGF-18

50 µG
Catalog # STMC78041.1
Human Recombinant FGF-18
Catalog # STMC78041.1
CAS Number:  
Human Recombinant FGF-18
Human Recombinant FGF-18

50 µG

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 50 µG
Product Details & Documents

Fibroblast growth factor 18 (FGF-18) is a growth factor and member of the FGF subfamily. FGF-18 is most similar to FGF-8 and FGF-17, and is secreted in adult lung and developing tissues (Ohbayashi et al.). FGF-18 signals through FGF receptor 3 (FGFR3) to regulate proliferation, differentiation, and matrix production of articular and growth plate chondrocytes in vivo and in vitro (Davidson et al.). FGF-18 has skeletal functions and protects articular cartilage by gene expression profiling and regulates proliferation and differentiation of midline cerebellar structures (Mori et al.). Also, recombinant human FGF-18 has been shown to effectively regulate hair growth (Song et al.).

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    50 µG
  • Cat. No.:
    STMC78041.1
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    FGF-18
  • Purity:
    ≥95%
  • Species:
    Human

Specifications