Order Entry
Export
Orders LinkContactUsLinkComponent
Mouse Recombinant IGF-I

Mouse Recombinant IGF-I

50 µG
Catalog # STMC78078.1
Mouse Recombinant IGF-I
Catalog # STMC78078.1
CAS Number:  
Mouse Recombinant IGF-I
Mouse Recombinant IGF-I

50 µG

CAS Number:  
Price
275,00 €
Log in for availability
1 * 50 µG
Product Details & Documents

Insulin-like growth factor 1 (IGF-I) is a polypeptide which belongs to the family of insulin-like growth factors that are similar in molecular structure to proinsulin. IGF-I binds to the IGF-I receptor and is a potent activator of the PI3K/AKT pathway; it also activates ERK1/2 signaling. IGF-I is required for embryonic development, and it is produced mainly in liver in response to a hepatocyte growth hormone. In the absence of insulin, IGF-I is necessary for the maintenance of human pluripotent stem cells (Wang et al.). Together with interleukin 3 (IL-3), IGF-I stimulates differentiation and proliferation of myeloid cells, and has been shown to regulate lymphopoiesis by stimulating proliferation and differentiation of T and B cells in lymphoid organs (Heemskerk et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    50 µG
  • Cat. No.:
    STMC78078.1
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IGF-I
  • Purity:
    ≥95%
  • Species:
    Mouse

Specifications