Order Entry
Export
Orders LinkContactUsLinkComponent
 

Mouse Recombinant IL-10

50 µG
Catalog # STMC78079.1
Mouse Recombinant IL-10
Catalog # STMC78079.1
CAS Number:  
Mouse Recombinant IL-10
Mouse Recombinant IL-10

50 µG

CAS Number:  
Price
802,00 €
Log in for availability
1 * 50 µG
Product Details & Documents

Interleukin 10 (IL-10) is the founding member of the IL-10 family of class II cytokines. All of the IL-10 cytokine family members have a four-helix bundle consisting of α-helical folds. Upon binding to its receptor, IL-10 activates signaling through JAK1 and STAT3. It is produced by dendritic cells, macrophages, and CD4+ T regulatory cells, as well as mast cells, NK cells, neutrophils, and regulatory B cells, under specific stimulating conditions (Saraiva and O'Garra). IL-10 can inhibit the activation of certain immune cells while it promotes the function of B cells, and facilitate healing process. Specifically, this cytokine is important for the function of T regulatory cells as it is a potent suppressor of effector T cell proliferation and cytokine production. Also, IL-10 produced by a subset of macrophages inhibits activation and production of pro-inflammatory cytokines by neighboring macrophages, thus allowing a level of self-regulation. IL-10 enhances B cell proliferation, immunoglobulin secretion, and class II MHC expression (Ouyang et al.).

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    50 µG
  • Cat. No.:
    STMC78079.1
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-10
  • Purity:
    ≥95 %
  • Species:
    Mouse

Specifications