Orders LinkContactUsLinkComponent
EasySep™ Human CD56 Positive Selection Kit II
EasySep™ Human CD56 Positive Selection Kit II
EasySep™ human CD56 positive selection kit II, 1×10⁹ cells
Catalog # STMC17855
EasySep™ Human CD56 Positive Selection Kit II
Catalog # STMC17855
EasySep™ Human CD56 Positive Selection Kit II
EasySep™ Human CD56 Positive Selection Kit II
EasySep™ human CD56 positive selection kit II, 1×10⁹ cells
Price
970,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Immunomagnetic positive selection of human CD56+ cells (e.g. NK cells).
  • Fast and easy-to-use
  • Up to 98% purity
  • No columns required
The EasySep™ Human CD56 Positive Selection Kit II is designed to isolate CD56+ cells from fresh or previously frozen peripheral blood mononuclear cells and human skeletal muscle (myoblasts and fibroblasts) cultures by positive selection. Desired cells are targeted with antibody complexes recognising CD56 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimise nonspecific binding. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD56 antigen is expressed on NK cells, NKT cells and human myoblasts.
This product replaces the EasySep™ Human CD56 Positive Selection Kit (STMC18055) for even faster cell isolations.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the ‘Product Restrictions’).
Specifications
  • Description:
    EasySep™ human CD56 positive selection kit II, 1×10⁹ cells
  • Seq:
    01