Order Entry
Export
Orders LinkContactUsLinkComponent
Human Recombinant IL-33

Human Recombinant IL-33

100 µG
Catalog # STMC78043
Human Recombinant IL-33
Catalog # STMC78043
CAS Number:  
Human Recombinant IL-33
Human Recombinant IL-33

100 µG

CAS Number:  
Price
1 330,00 €
Log in for availability
1 * 100 µG
Product Details & Documents

Interleukin 33 (IL-33) is a pro-inflammatory cytokine from the IL-1 family. It binds to ST2 receptor and activates NF-κB and MAPK pathways. IL-33 is expressed by epithelial cells, smooth muscle cells, and fibroblasts in various tissues and organs, as well as resting basophils, mast cells, eosinophils, natural helper cells, group 2 innate lymphoid cells, dendritic cells, and activated macrophages (Schmitz et al.; Yasuda et al.). It contributes to allergic inflammation by stimulating production of the cytokines IL-4, IL-5, and IL-13, and stimulates host defense against microbial and viral infections (Liew; Yasuda et al.). In the central nervous system, IL-33 is produced by endothelial cells and astrocytes. It induces proliferation of microglia and mediates production of pro-inflammatory cytokines (Yasuoka et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    100 µG
  • Cat. No.:
    STMC78043
  • Protein/peptide type:
    Recombinant
  • Protein/peptide name:
    IL-33
  • Species:
    Human

Specifications