Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Low-Tox thermometer brown spirit, total immersion, -10 to +330C, 305mm 1 * 1 items
Catalog # BRNN44/827/0
Supplier:
S BRANNAN AND SONS
Expand image
Low-Tox thermometer brown spirit, total immersion, -10 to +330C, 305mm 1 * 1 items
Catalog # BRNN44/827/0
Supplier:
S BRANNAN AND SONS