ContactUsLinkComponent
SAPPHIRE INJECTOR. HF-RESISTANT. THESE SAPPHIRE INJECTORS ARE USED FOR ENVIRONMENTAL, GEOCHEMISTRY, AND HF APPLICATIONS ON THE OPTIM A 8X00 SERIES INSTRUMENTS. PLEASE NOTE: COMPATIBLE WITH ESI PRESS-FIT PFA SPRAY CHAMBER CONNECTION. 1 * 1 items
Catalog # PERCN0782005
Supplier:  PerkinElmer
SAPPHIRE INJECTOR. HF-RESISTANT. THESE SAPPHIRE INJECTORS ARE USED FOR ENVIRONMENTAL, GEOCHEMISTRY, AND HF APPLICATIONS ON THE OPTIM A 8X00 SERIES INSTRUMENTS. PLEASE NOTE: COMPATIBLE WITH ESI PRESS-FIT PFA SPRAY CHAMBER CONNECTION. 1 * 1 items
Catalog # PERCN0782005
Supplier:  PerkinElmer