pH buffers, ready to use
Technical buffer TPL 10 Trace/25, 10,01, Plastic bottle, 25
Catalog # WTWA108814
Supplier: WTW
Jump to:
Product Details & Documents
Every bottle has its own lot reference with the expiration date.
- Shelf life up to 3 years (unopened)
- Reference temperature at 25 °C
- Economic consumption
- Easiest handling by use of the dosing vessel
- One litre container with a separate compartment for the sample
Accuracy is ±0,03 pH
: Traceable to NIST.
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:25
- Description:Technical buffer TPL 10 Trace/25
- pH value:10,01



