Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
FAGOMINE
FAGOMINE
Print
FAGOMINE
Catalog # MDEXHY-13005-5MG
Supplier:
MEDCHEMEXPRESS
FAGOMINE
Catalog # MDEXHY-13005-5MG
Supplier:
MEDCHEMEXPRESS
CAS Number:
Expand image
Return to Top
FAGOMINE
Add to cart
CAS Number:
Price
438,00 €
Log in
for availability
Unit of Measure
1 * 5 mg
-
+
Add to cart
Return Policy
Product Details & Documents