Orders LinkContactUsLinkComponent
Shoe covers, Secutex® pro, PP/PE
Shoe covers, Secutex® pro, PP/PE
With reinforced sole, White, U*
Catalog # ZVGA30254-90
Shoe covers, Secutex® pro, PP/PE
Catalog # ZVGA30254-90
Shoe covers, Secutex® pro, PP/PE
Shoe covers, Secutex® pro, PP/PE
With reinforced sole, White, U*
Price
47,60 €
Log in for availability
1 * 25 PAIR
Product Details & Documents
These shoe covers protect against aggressive chemicals and other substances.
  • Lint-free, resistant to abrasion, tear and water
  • Permeable to air and water vapour ensures breathability
  • Anti-static
*One size fits all.
Documents
Specifications
  • Shoe size:
    U*
  • Colour:
    White
  • Description:
    With reinforced sole
  • Cat. No.:
    ZVGA30254-90
  • Garment application:
    Chemical protection|Laboratory