Shoe covers, Secutex® pro, PP/PE
Standard, White, U*
Catalog # ZVGA30250-90
Supplier: Zellstoff-Vertriebs-GmbH & Co. KG
Jump to:
Product Details & Documents
These shoe covers protect against aggressive chemicals and other substances.
- Lint-free, resistant to abrasion, tear and water
- Permeable to air and water vapour ensures breathability
- Anti-static
*One size fits all.
Documents
ZVGA30254-90_TDS_EN
Datasheet
Specifications
- Shoe size:U*
- Colour:White
- Description:Standard
- Cat. No.:ZVGA30250-90
- Garment application:Chemical protection|Laboratory